Anti-Casein Kinase 2 beta Goat Polyclonal Antibody
Catalog # 77063-310
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Casein Kinase 2 beta
- Antigen Symbol:Casein Kinase 2 beta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Pig,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:Casein kinase II subunit beta|CK2B|CK II beta|CSNK 2B|polypeptide|Casein kinase II beta subunit|CSK2B|CSK2B_HUMAN|CK2N
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-TGLNEQVPHYRQ
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human CSNK2B (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77063-310
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to Casein Kinase 2 beta for ELISA and WB with samples derived from Human, Mouse, Rat and Porcine.
- Validated Applications: ELISA, WB
Type: Primary
Antigen: Casein Kinase 2 beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine