Anti-BCL7A Goat Polyclonal Antibody
Catalog # 77067-526
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:B cell CLL/lymphoma 7
- Antigen Symbol:BCL7A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:BCL7A_HUMAN|bcl7a|A|Bcl 7A|B-cell CLL/lymphoma 7 protein family member A|BCL 7|protein family member A|BCL7
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-MKLEASQQNSEEM
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human BCL7A (C terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77067-526
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to BCL7A for ELISA, WB, IF and Flow Cytometry with samples derived from Human.
- Validated Applications: ELISA, WB, IF, Flow Cytometry
Type: Primary
Antigen: BCL7A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human