Anti-Desmoplakin Goat Polyclonal Antibody
Catalog # 77078-520
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:250/210 kDa paraneoplastic pemphigus antigen
- Antigen Symbol:Desmoplakin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:DP I|Desmoplakin|KPPS2|DPI|DSP|Desmoplakin I|DP II|Desmoplakin II|DPII
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-QESVKQAHLRNEH
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human DSP (internal region)
- Tested Applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77078-520
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to Desmoplakin for ELISA and IHC with samples derived from Human.
- Validated Applications: ELISA, IHC
Type: Primary
Antigen: Desmoplakin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human