Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Syntaxin 8 Rabbit Polyclonal Antibody
Anti-Syntaxin 8 Rabbit Polyclonal Antibody
Supplier:  ANTIBODIES.COM LLC
Certificates
undefined
Anti-Syntaxin 8 Rabbit Polyclonal Antibody
Supplier:  ANTIBODIES.COM LLC

Specifications

  • Catalog No:
  • 76850-884
  • Antigen Symbol:
  • Syntaxin 8
  • Antigen Name:
  • CARB
  • Antigen Synonyms:
  • STX 8|Syntaxin8|STX8_HUMAN|Stx8|Syntaxin 8|Syntaxin-8
  • Antibody Type:
  • Primary
  • Clonality:
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Reactivity:
  • Human, Mouse
  • Host:
  • Rabbit
  • Gene ID:
  • UniprotID# Q9UNK0
  • Isotype:
  • IgG
  • Western Blot:
  • Yes
  • Immunogen:
  • Recombinant fusion protein containing a sequence corresponding to amino acids 1 - 215 of human STX8 (NP_004844.1)
  • Sequence:
  • MAPDPWFSTYDSTCQIAQEIAEKIQQRNQYERKGEKAPKLTVTIRALLQNLKEKIALLKDLLLRAVSTHQITQLEGDRRQNLLDDLVTRERLLLASFKNEGAEPDLIRSSLMSEEAKRGAPNPWLFEEPEETRGLGFDEIRQQQQKIIQEQDAGLDALSSIISRQKQMGQEIGNELDEQNEIIDDLANLVENTDEKLRNETRRVNMVDRKSASCG
  • Form:
  • Liquid
  • Molecular Weight:
  • 27 kDa
  • Purification:
  • Affinity purification
  • Storage Buffer:
  • Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.02% Sodium Azide
  • Storage Temperature:
  • Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping Temperature:
  • Shipped on blue ice at 4 °C

Specifications

Related Information