About this item
Rabbit polyclonal antibody to Met (c-Met) (phospho Tyr1234 + Tyr1235) for WB with samples derived from Human.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1000
Type: Primary
Antigen: Met (c-Met)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77217-004
- Antigen Symbol:
- Met (c-Met)
- Antigen Name:
- AUTS9
- Antigen Synonyms:
- HGFR|Met proto oncogene tyrosine kinase|c met|HGF/SF receptor|Hepatocyte growth factor receptor|HGF|D249|Met (c-Met)|HGF receptor
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- UniprotID# P08581
- Isotype:
- IgG
- Western Blot:
- Yes
- Modification:
- phospho Tyr1234 + Tyr1235
- Immunogen:
- A synthetic phosphorylated peptide around Y1234 and Y1235 of human MET (NP_000236.2)
- Sequence:
- KEYYSV
- Form:
- Liquid
- Molecular Weight:
- 145 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.05% Proclin 300
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C