Rabbit monoclonal [ARC0107] antibody to S6K1 (phospho Thr421 + Ser424) for WB, IHC and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ICC/IF
- Recommended Dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: S6K1
Clonality: Monoclonal
Clone: ARC0107
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77217-000
- Antigen Symbol:
- S6K1
- Antigen Name:
- 70 kDa ribosomal protein S6 kinase 1
- Antigen Synonyms:
- P70 beta 1|KS6B1_HUMAN|P70 S6 Kinase|p70 alpha|p70 S6 kinase, alpha 2|p70 S6 kinase alpha|p70 ribosomal S6 kinase alpha|p70 S6 kinase, alpha 1|p70 ribosomal S6 kinase beta 1
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC0107
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# P23443
- Isotype:
- IgG
- Western Blot:
- Yes
- ImmunoChemistry:
- Yes
- Modification:
- phospho Thr421 + Ser424
- Immunogen:
- A synthesized peptide derived from human Phospho-S6K1 (T421 + S424)
- Form:
- Liquid
- Molecular Weight:
- 70 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C
- Tested Applications:
- IHC, ICC/IF