Anti-PLA2G7 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to PLA2G7 for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1000
Type: Primary
Antigen: PLA2G7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 76857-518
- Antigen Symbol:
- PLA2G7
- Antigen Name:
- 1 alkyl 2 acetylglycerophosphocholine esterase
- Antigen Synonyms:
- 2-acetyl-1-alkylglycerophosphocholine esterase|gVIIA PLA2|Group VIIA phospholipase A2|gVIIA-PLA2|LDL PLA(2)|2 acetyl 1 alkylglycerophosphocholine esterase|1-alkyl-2-acetylglycerophosphocholine esterase|EC 3.1.1.47 PAF acetylhydrolase PAF 2-acylhydrolase LDL associated phospholipase A2|Group-VIIA phospholipase A2
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q13093
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 172 - 441 of human PLA2G7 (NP_005075.3)
- Sequence:
- FIVAAVEHRDRSASATYYFKDQSAAEIGDKSWLYLRTLKQEEETHIRNEQVRQRAKECSQALSLILDIDHGKPVKNALDLKFDMEQLKDSIDREKIAVIGHSFGGATVIQTLSEDQRFRCGIALDAWMFPLGDEVYSRIPQPLFFINSEYFQYPANIIKMKKCYSPDKERKMITIRGSVHQNFADFTFATGKIIGHMLKLKGDIDSNVAIDLSNKASLAFLQKHLGLHKDFDQWDCLIEGDDENLIPGTNINTTNQHIMLQNSSGIEKYN
- Form:
- Liquid
- Molecular Weight:
- 60 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C