Anti-GLUT3 Rat Monoclonal Antibody [clone: #10]
Catalog # 77069-494
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Symbol:GLUT3
- Clonality:Monoclonal
- Clone:#10
- Conjugation:Unconjugated
- Host:Rat
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P32037
- Storage Buffer:Supplied in Phosphate Buffered Saline with 50% Glycerol (filter sterilised, without Sodium Azide, and carrier free)
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Mouse sperm
- Purification:Affinity purification
- Cat. No.:77069-494
- Packaging:Plastic vial
Specifications
About this item
Rat monoclonal (#10) antibody to GLUT3 for WB with samples derived from Mouse.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:200 to 1:500
Type: Primary
Antigen: GLUT3
Clonality: Monoclonal
Clone: #10
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Mouse