Anti-GAK Mouse Monoclonal Antibody [clone: 9-10]
Catalog # 77077-880
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Auxilin 2
- Antigen Symbol:GAK
- Clonality:Monoclonal
- Clone:9-10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:UniprotID# P97874
- Antigen Synonyms:DNAJC26|Cyclin G associated kinase|EC 2.7.11.1|GAK protein|Cyclin-G-associated kinase|FLJ40395|FLJ16629|DNAJ26|GAK
- Storage Buffer:Supplied in Phosphate Buffered Saline with 50% Glycerol
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant rat GAK protein (a.a. 1-430)
- Tested Applications:IHC
- Cat. No.:77077-880
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal (9-10) antibody to GAK for WB, IF and IHC with samples derived from Human and Rat.
- Validated Applications: WB, IF, IHC
- Recommended Dilutions: WB: 1:500, IF: 1:50, IHC: 1:50
Type: Primary
Antigen: GAK
Clonality: Monoclonal
Clone: 9-10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat