To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Transcriptional adaptor 3 like
- Antigen Symbol:ADA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:S. cerevisiae
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:UniprotID# P32494
- Antigen Synonyms:ADA3|ADA3-like protein|TADA3L|hADA3|FLJ20221|ADA3 homolog|FLJ21329|Transcriptional adaptor 3 (NGG1 homolog yeast) like
- Storage Buffer:Supplied as antiserum with 0.1% Sodium Azide
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant ADA3 protein (His tag), produced in E. coli
- Cat. No.:77077-736
- Packaging:Plastic vial
Rabbit polyclonal antibody to ADA3 for WB with samples derived from Saccharomyces cerevisiae.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1000
Type: Primary
Antigen: ADA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Saccharomyces cerevisiae