Anti-CCR8 Recombinant Monoclonal Antibody [clone: BMS 986340]
Catalog # 77187-948
Supplier: Antibodies.com
Specifications
- Antibody Type:Primary
- Antigen Name:C C chemokine receptor type 8
- Antigen Symbol:CCR8
- Clonality:Recombinant Monoclonal
- Clone:BMS 986340
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Chimera
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Form:Lyophilized
- Gene ID:UniprotID# P51685
- Antigen Synonyms:C-C CKR-8|CC chemokine receptor CHEMR1|C C CKR 8|CC CKR 8|CC chemokine receptor 8|C-C chemokine receptor type 8|C C motif chemokine receptor 8|CC chemokine receptor type 8|CC-CKR-8
- Storage Buffer:Lyophilized from sterile phosphate buffered saline, pH 7.4. Normally 5 - 8% trehalose is added as a protectant before lyophilization
- Storage Temperature:Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Concentration:Reconstitution dependent
- Shipping Temperature:Shipped on blue ice at 4 °C
- Cat. No.:77187-948
- Packaging:Plastic vial
Specifications
About this item
Recombinant humanized monoclonal [BMS 986340] antibody to CCR8 for ELISA and flow cytometry with samples derived from human.
- Validated applications: ELISA, Flow cytometry
- Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100
Type: Primary
Antigen: CCR8
Clonality: Recombinant
Clone: BMS 986340
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human