Anti-CD96 Chimera Recombinant Monoclonal Antibody [clone: DMC285]
Catalog # 77187-730
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:CD96
- Antigen Symbol:CD96
- Clonality:Recombinant Monoclonal
- Clone:DMC285
- Conjugation:Unconjugated
- Host:Chimera
- Isotype:IgG1
- Reactivity:Human
- Form:Lyophilized
- Antigen Synonyms:CD96 molecule|Cell surface antigen CD96|DKFZp667E2122|MGC22596|T cell activated increased late expression protein|T cell activation, increased late expression|T cell surface protein tactile precursor|T cell-activated increased late expression protein|T-cell surface protein tactile
- Storage Buffer:Lyophilized from sterile phosphate buffered saline, pH 7.4. Normally 5 - 8% trehalose is added as a protectant before lyophilization
- Storage Temperature:Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Cat. No.:77187-730
Specifications
About this item
Recombinant chimeric monoclonal [DMC285] antibody to CD96 for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1:100
Type: Primary
Antigen: CD96
Clonality: Recombinant
Clone: DMC285
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human