Anti-EMR2 Chimera Recombinant Monoclonal Antibody [clone: DMC370]
Catalog # 77187-690
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:CD 312
- Antigen Symbol:EMR2
- Clonality:Recombinant Monoclonal
- Clone:DMC370
- Conjugation:Unconjugated
- Host:Chimera
- Isotype:IgG1
- Reactivity:Human
- Form:Lyophilized
- Antigen Synonyms:CD 312 antigen|CD312|CD312 antigen|Egf like module containing mucin like hormone receptor like 2|Egf like module containing mucin like hormone receptor like sequence 2|EGF like module EMR2|EGF-like module receptor 2|EGF-like module-containing mucin-like hormone receptor-like 2|EMR2_HUMAN
- Storage Buffer:Lyophilized from sterile phosphate buffered saline, pH 7.4. Normally 5 - 8% trehalose is added as a protectant before lyophilization
- Storage Temperature:Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Cat. No.:77187-690
Specifications
About this item
Recombinant chimeric monoclonal [DMC370] antibody to EMR2 for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1:100
Type: Primary
Antigen: EMR2
Clonality: Recombinant
Clone: DMC370
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human