Anti-CLEC1 Chimera Recombinant Monoclonal Antibody [clone: DMC468]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Recombinant chimeric monoclonal [DMC468] antibody to CLEC1 for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1:100
Type: Primary
Antigen: CLEC1
Clonality: Recombinant
Clone: DMC468
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- 77187-528
- 77187-526
- Antigen Symbol:
- CLEC1
- CLEC1
- Antigen Name:
- 5930406N14Rik
- 5930406N14Rik
- Antigen Synonyms:
- MGC34328|OTTHUMP00000238867|CLC1A_HUMAN|C-type lectin-like receptor 1|OTTHUMP00000238868|CLEC1A|CLEC-1|C-type lectin domain family 1 member A|CLEC1
- MGC34328|OTTHUMP00000238867|CLC1A_HUMAN|C-type lectin-like receptor 1|OTTHUMP00000238868|CLEC1A|CLEC-1|C-type lectin domain family 1 member A|CLEC1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Recombinant Monoclonal
- Recombinant Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- DMC468
- DMC468
- Reactivity:
- Human
- Human
- Host:
- Chimera
- Chimera
- Gene ID:
- UniprotID# Q8NC01
- UniprotID# Q8NC01
- Isotype:
- IgG1
- IgG1
- Flow Cytometry:
- Yes
- Yes
- Form:
- Lyophilized
- Lyophilized
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- Reconstitution dependent
- Reconstitution dependent
- Storage Buffer:
- Lyophilized from sterile phosphate buffered saline, pH 7.4. Normally 5 - 8% trehalose is added as a protectant before lyophilization
- Lyophilized from sterile phosphate buffered saline, pH 7.4. Normally 5 - 8% trehalose is added as a protectant before lyophilization
- Storage Temperature:
- Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C