To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Glycoprotein 75
- Antigen Symbol:TRP1
- Clonality:Recombinant
- Clone:TYRP1/2340R
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P17643
- Antigen Synonyms:CAS2|Catalase B|6-dihydroxyindole-2-carboxylic acid oxidase|GP75|CATB|b-PROTEIN|5,6 dihydroxyindole 2 carboxylic acid oxidase|DHICA oxidase|5
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:75 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant fragment, within amino acids 257 - 377, of human TYRP1 protein. The exact sequence is proprietary
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76951-550
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [TYRP1/2340R] antibody to TRP1 for ELISA and IHC-P with samples derived from Human.
- Validated Applications: ELISA, IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: TRP1
Clonality: Recombinant
Clone: TYRP1/2340R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human