Anti-EZH2 Mouse Monoclonal Antibody [clone: EZH2/2536]
Catalog # 76962-028
Supplier: Antibodies.com
Specifications
- Antibody Type:Primary
- Antigen Name:Enhancer of zeste 2
- Antigen Symbol:EZH2
- Clonality:Monoclonal
- Clone:EZH2/2536
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q15910
- Antigen Synonyms:Enhancer of zeste homolog 2 (Drosophila)|enhancer of zeste 2 polycomb repressive complex 2 subunit|Enx1h|ENX1|Enhancer of zeste, Drosophila, homolog 2|Enhancer of zeste homolog 2|ENX-1|Enx 1h|ENX 1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:81 - 102 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant full-length human EZH2 protein
- Purification:Protein A/G chromatography
- Cat. No.:76962-028
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [EZH2/2536] antibody to EZH2 for WB with samples derived from Human.
- Validated Applications: WB
- Recommended Dilutions: WB: 1 to 2 µg/ml
Type: Primary
Antigen: EZH2
Clonality: Monoclonal
Clone: EZH2/2536
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human