Anti-CD34 Rabbit Recombinant Antibody [clone: HPCA1/2598R]
Catalog # 76965-760
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:CD34
- Antigen Symbol:CD34
- Clonality:Recombinant
- Clone:HPCA1/2598R
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P28906
- Antigen Synonyms:CD34 antigen|Cluster designation 34|Hematopoietic progenitor cell antigen CD34|CD34_HUMAN|Mucosialin|CD34 molecule|OTTHUMP00000034734|HPCA1|OTTHUMP00000034733
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:90 - 110 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant full-length human HPCA1 protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76965-760
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [HPCA1/2598R] antibody to CD34 for Flow Cytometry and IHC-P with samples derived from Human and Rat.
- Validated Applications: Flow Cytometry, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD34
Clonality: Recombinant
Clone: HPCA1/2598R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat