Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-TIF1 alpha Mouse Monoclonal Antibody [clone: PCRP-TRIM24-1B12]
Anti-TIF1 alpha Mouse Monoclonal Antibody [clone: PCRP-TRIM24-1B12]
Catalog # 76953-178
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-TIF1 alpha Mouse Monoclonal Antibody [clone: PCRP-TRIM24-1B12]
Catalog # 76953-178
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A250442-100

Specifications

  • Size:
    100 µg
  • Cat. No.:
    76953-178

Specifications

About this item

Mouse monoclonal [PCRP-TRIM24-1B12] antibody to TIF1 alpha for Flow Cytometry and IF with samples derived from Human.

  • Validated Applications: Flow Cytometry, IF
  • Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml

Type: Primary
Antigen: TIF1 alpha
Clonality: Monoclonal
Clone: PCRP-TRIM24-1B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human