Anti-TRP1 Mouse Monoclonal Antibody [clone: TYRP1/3280]
76956-626
- Antibody Type:Primary
- Antigen Name:GP75
- Antigen Symbol:TRP1
- Clonality:Monoclonal
- Clone:TYRP1/3280
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P17643
- Antigen Synonyms:CAS2|Catalase B|6-dihydroxyindole-2-carboxylic acid oxidase|CATB|Glycoprotein 75|b-PROTEIN|5,6 dihydroxyindole 2 carboxylic acid oxidase|DHICA oxidase|5
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:75 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant fragment, within amino acids 257 - 377, of human TYRP1 protein. The exact sequence is proprietary
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76956-626
- Packaging:Plastic vial
Mouse monoclonal [TYRP1/3280] antibody to TRP1 for ELISA and IHC-P with samples derived from Human.
- Validated Applications: ELISA, IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: TRP1
Clonality: Monoclonal
Clone: TYRP1/3280
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human