Anti-Mucin 5AC Mouse Monoclonal Antibody [clone: 2-11M1]
Catalog # 76962-846
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Apomucin
- Antigen Symbol:Mucin 5AC
- Clonality:Monoclonal
- Clone:2-11M1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Monkey,Human,Bovine,Mouse,Cat
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P98088
- Antigen Synonyms:Lewis B blood group antigen|Gastric mucin|Mucin 5 Subtypes A And C|MUC5AC|Apomucin Major Airway Glycoprotein|MUC 5|MUC5|Mucin 5 subtype AC tracheobronchial|LeB
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:>1000 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b) patient
- Purification:Protein A/G chromatography
- Cat. No.:76962-846
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [2-11M1] antibody to Mucin 5AC for ELISA, Flow Cytometry and IF with samples derived from Human, Monkey, Bovine, Feline and Mouse.
- Validated Applications: ELISA, Flow Cytometry, IF
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: Mucin 5AC
Clonality: Monoclonal
Clone: 2-11M1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Monkey;Bovine;Cat;Mouse