Anti-MUC1 Mouse Monoclonal Antibody [clone: MUC1/520]
76954-558
- Size:100 µg
- Cat. No.:76954-558
Mouse monoclonal [MUC1/520] antibody to MUC1 for IHC with samples derived from Human.
- Validated Applications: IHC
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: MUC1/520
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a lambda
Reactivity: Human