Anti-Mammaglobin A Mouse Monoclonal Antibody [clone: MGB/2704]
76950-834
- Size:100 µg
- Cat. No.:76950-834
Mouse monoclonal [MGB/2704] antibody to Mammaglobin A for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Mammaglobin A
Clonality: Monoclonal
Clone: MGB/2704
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human