Anti-Mammaglobin A Mouse Monoclonal Antibody [clone: MGB/2000]
76960-460
- Size:100 µg
- Cat. No.:76960-460
Mouse monoclonal [MGB/2000] antibody to Mammaglobin A for WB and IHC-P with samples derived from Human.
- Validated Applications: WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Mammaglobin A
Clonality: Monoclonal
Clone: MGB/2000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human