Anti-Milk Fat Globule Mouse Monoclonal Antibody [clone: EDM45]
76960-644
- Size:100 µg
- Cat. No.:76960-644
Mouse monoclonal [EDM45] antibody to Milk Fat Globule for Flow Cytometry, IF and IHC-P with samples derived from Human.
- Validated Applications: Flow Cytometry, IF, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Milk Fat Globule
Clonality: Monoclonal
Clone: EDM45
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human