Anti-c-Kit Mouse Monoclonal Antibody [clone: KIT/2669]
76962-952
- Antibody Type:Primary
- Antigen Name:C Kit
- Antigen Symbol:c-Kit
- Clonality:Monoclonal
- Clone:KIT/2669
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P10721
- Antigen Synonyms:c-Kit Ligand|KIT oncogene|KIT_HUMAN|KIT proto oncogene receptor tyrosine kinase|CD117|Mast cell growth factor receptor|c-Kit|Mast/stem cell growth factor receptor precursor|Mast/stem cell growth factor receptor Kit
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:~145 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant full-length human KIT protein
- Purification:Protein A/G chromatography
- Cat. No.:76962-952
- Packaging:Plastic vial
Mouse monoclonal [KIT/2669] antibody to c-Kit for ELISA with samples derived from Human.
- Validated Applications: ELISA
Type: Primary
Antigen: c-Kit
Clonality: Monoclonal
Clone: KIT/2669
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human