To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Cadherin family member 7
- Antigen Symbol:FAT1
- Clonality:Monoclonal
- Clone:FAT1-3D7/1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgM kappa
- Reactivity:Drosophila melanogaster
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q14517
- Antigen Synonyms:CDHF 7|Cadherin ME5|Cadherin related tumor suppressor homolog|Cadherin related tumor suppressor|Cadherin family member 7 precursor|Cadherin-related tumor suppressor homolog|CDHF7|Cadherin related tumor suppressor homolog precursor|Cadherin related tumor suppressor homolog precursor (FAT protein homolog)
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:500 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Cytoplasmic domain of drosophila FAT protein
- Purification:Protein A/G chromatography
- Cat. No.:76955-810
- Packaging:Plastic vial
Mouse monoclonal [FAT1-3D7/1] antibody to FAT1 for ELISA, WB and IF with samples derived from Drosophila melanogaster.
- Validated Applications: ELISA, WB, IF
- Recommended Dilutions: WB: 1 to 2 µg/ml, IF: 1 to 2 µg/ml
Type: Primary
Antigen: FAT1
Clonality: Monoclonal
Clone: FAT1-3D7/1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Drosophila melanogaster