Anti-EIF4A2 Mouse Monoclonal Antibody [clone: PCRP-EIF4A2-2B5]
Catalog # 76964-314
Supplier: Antibodies.com
Specifications
- Antibody Type:Primary
- Antigen Name:ATP dependent RNA helicase eIF4A 2
- Antigen Symbol:EIF4A2
- Clonality:Monoclonal
- Clone:PCRP-EIF4A2-2B5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q14240
- Antigen Synonyms:eIF4A-II|eIF 4A II|EIF4A|eIF4A2|eIF4A II|BM-010|ATP-dependent RNA helicase eIF4A-2|eIF-4A-II|DDX2B
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:46 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant full-length human EIF4A2 protein
- Purification:Protein A/G chromatography
- Cat. No.:76964-314
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PCRP-EIF4A2-2B5] antibody to EIF4A2 for ELISA with samples derived from Human.
- Validated Applications: ELISA
Type: Primary
Antigen: EIF4A2
Clonality: Monoclonal
Clone: PCRP-EIF4A2-2B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human