To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Colony stimulating factor
- Antigen Symbol:GM-CSF
- Clonality:Monoclonal
- Clone:SPM469
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rat
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Cynomolgus monkey,Rhesus monkey,Human
- Size:100 µg
- Environmentally Preferable:
- Neutralisation:Yes
- Form:Liquid
- Gene ID:UniprotID# P04141
- Antigen Synonyms:CSF2_HUMAN|GMCSF|Colony-stimulating factor|Colony Stimulating Factor 2|GM-CSF|Colony stimulating factor 2 (granulocyte-macrophage)|CSF2|CSF 2|CSF
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:22 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant human GM-CSF protein
- Purification:Protein A/G chromatography
- Cat. No.:76962-492
- Packaging:Plastic vial
Rat monoclonal [SPM469] antibody to GM-CSF for Neutralization Studies, Flow Cytometry and IF with samples derived from Human, Cynomolgus Monkey and Rhesus Monkey.
- Validated Applications: Neutralization Studies, Flow Cytometry, IF
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: GM-CSF
Clonality: Monoclonal
Clone: SPM469
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human;Cynomolgus Monkey;Rhesus Monkey