To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:antigen MSK39 identified by monoclonal
- Antigen Symbol:CD56
- Clonality:Monoclonal
- Clone:LT56
- Conjugation:PE (Phycoerythrin)-Cy7®
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Size:100 tests
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P13591
- Antigen Synonyms:NCAM 1|NCAM|N-CAM-1|MSK39|CD56|antigen recognized by monoclonal|cell adhesion molecule, neural, 1|MSK 39|NCAM C
- Storage Buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage Temperature:Store in the dark at 2...8 ℃. Do not freeze!
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Cell line KG1a
- Purification:This antibody is conjugated with tandem dye PE-Cyanine 7 under optimum conditions The conjugate is purified by size-exclusion chromatography
- Cat. No.:76967-410
- Packaging:Glass bottle
Specifications
About this item
Mouse monoclonal [LT56] antibody to CD56 (PE-Cyanine 7) for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD56
Clonality: Monoclonal
Clone: LT56
Conjugation: PE-Cyanine 7
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human