To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CD34
- Antigen Symbol:CD34
- Clonality:Monoclonal
- Clone:581
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5,5®
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Non-human primates,Human
- Size:100 tests
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P28906
- Antigen Synonyms:CD34 antigen|Cluster designation 34|Hematopoietic progenitor cell antigen CD34|CD34_HUMAN|Mucosialin|CD34 molecule|OTTHUMP00000034734|HPCA1|OTTHUMP00000034733
- Storage Buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage Temperature:Store in the dark at 2...8 ℃. Do not freeze!
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Unfortunately, this information is considered to be co mMercially sensitive
- Purification:This antibody is conjugated with tandem dye PerCP-Cyanine 55 under optimum conditions The conjugate is purified by size-exclusion chromatography
- Cat. No.:76962-882
- Packaging:Glass bottle
Specifications
About this item
Mouse monoclonal [581] antibody to CD34 (PerCP-Cyanine 5.5) for Flow Cytometry with samples derived from Human and Non-Human Primates.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 581
Conjugation: PerCP-Cyanine 5.5
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Non-Human Primates