To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Cell surface glycoprotein Trop 2
- Antigen Symbol:CD326
- Clonality:Monoclonal
- Clone:323/A3
- Conjugation:APC (Allophycocyanin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 tests
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P16422
- Antigen Synonyms:CD326|Antigen identified by monoclonal|Cell surface glycoprotein Trop 1|AUA1|CD326 antigen|Adenocarcinoma-associated antigen|Adenocarcinoma associated antigen|323/A3|17 1A
- Storage Buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage Temperature:Store in the dark at 2...8 ℃. Do not freeze!
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Human breast cancer MCF-7 cells
- Purification:This antibody is conjugated with APC under optimum conditions The conjugate is purified by size-exclusion chromatography
- Cat. No.:76959-674
- Packaging:Glass bottle
Mouse monoclonal [323/A3] antibody to CD326 (APC) for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD326
Clonality: Monoclonal
Clone: 323/A3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human