To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:HLA ABCE
- Clonality:Monoclonal
- Clone:TP25.99SF
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Storage Buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Interferon ga mMa treated human melanoma cell line Colo 38
- Tested Applications:IHC-Fr
- Purification:Protein A chromatography
- Cat. No.:76955-484
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [TP25.99SF] antibody to HLA ABCE for Flow Cytometry, WB and IHC-Fr with samples derived from Human.
- Validated Applications: Flow Cytometry, WB, IHC to Fr
- Recommended Dilutions: Flow Cytometry: 1 to 4 µg/ml
Type: Primary
Antigen: HLA ABCE
Clonality: Monoclonal
Clone: TP25.99SF
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human