Anti-OAS2 Rabbit Polyclonal Antibody
Catalog # 77080-860
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:(2-5'')oligo(A) synthase 2
- Antigen Symbol:OAS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P29728
- Antigen Synonyms:2'-5'-oligoadenylate synthetase 2|(2-5')oligo(A) synthetase 2|2-prime, 5-prime-oligoadenylate synthetase 2 - 2-prime, 5-prime-oligoadenylate synthetase 2, 69-KD|2'-5'-oligoadenylate synthetase 2, 69/71kDa|2''-5''-oligoadenylate synthase 2|2-5A synthase 2|2-prime, 5-prime-oligoadenylate synthetase 2|MGC78578|2-5A synthetase 2
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:82 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from the N - terminal region of human OAS2 (amino acids 61 - 110)
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
- Cat. No.:77080-860
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to OAS2 for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: OAS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human