Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Thrombopoietin Rabbit Polyclonal Antibody
Anti-Thrombopoietin Rabbit Polyclonal Antibody
Catalog # 77065-818
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-Thrombopoietin Rabbit Polyclonal Antibody
Catalog # 77065-818
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A101289-100

Specifications

  • Size:
    100 µg
  • Cat. No.:
    77065-818

Specifications

About this item

Rabbit polyclonal antibody to THPO for WB and ELISA with samples derived from Human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: Thrombopoietin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human