Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Vesicle docking protein p115 Rabbit Polyclonal Antibody
Anti-Vesicle docking protein p115 Rabbit Polyclonal Antibody
Catalog # 77072-128
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-Vesicle docking protein p115 Rabbit Polyclonal Antibody
Catalog # 77072-128
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A101283-100

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    General vesicular transport factor
  • Antigen Symbol:
    Vesicle docking protein p115
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Form:
    Liquid
  • Gene ID:
    UniprotID# O60763
  • Antigen Synonyms:
    Vesicle docking protein|Protein USO1 homolog|VDP|Uso1|Transcytosis associated protein|Transcytosis-associated protein|USO1_HUMAN|General vesicular transport factor p115|TAP
  • Storage Buffer:
    Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
  • Molecular Weight:
    107 kDa
  • Storage Temperature:
    Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
  • Shipping Temperature:
    Shipped on blue ice at 4 ℃
  • Immunogen:
    Synthetic peptide derived from human USO1 (amino acids 913 - 962)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
  • Cat. No.:
    77072-128
  • Packaging:
    Plastic vial

Specifications

About this item

Rabbit polyclonal antibody to USO1 for WB and ELISA with samples derived from Human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000

Type: Primary
Antigen: Vesicle docking protein p115
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human