Anti-CLCC1 Rabbit Polyclonal Antibody
100 µg, Plastic vial
Catalog # 77076-034
Supplier: Antibodies.com
Supplier #: A101145-100
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Rabbit polyclonal antibody to CLCC1 for WB and ELISA with samples derived from Human.
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Type: Primary
Antigen: CLCC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Documents
Specifications
- Size:100 µg
- Packaging:Plastic vial
- Clonality:Polyclonal
- Molecular Weight:62 kDa
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Form:Liquid
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Isotype:IgG
- Host:Rabbit
- Cat. No.:77076-034
- Reactivity:Human
- Western Blot:Yes
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Antibody Type:Primary
- Antigen Symbol:CLCC1
- Gene ID:UniprotID# Q96S66
- Immunogen:Synthetic peptide derived from human CLCC1 (amino acids 380 - 429)
- Conjugation:Unconjugated
- Antigen Synonyms:Mid 1 related chloride channel 1|OTTMUSP00000007701|KIAA0761|Mid 1 related chloride channel protein 1|OTTMUSP00000007700|MCLC|RP23-217I18.1|CLCC1
- Antigen Name:Chloride channel CLIC like protein 1
- ELISA:Yes
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



