Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ICAM2 Rabbit Polyclonal Antibody
Anti-ICAM2 Rabbit Polyclonal Antibody
Catalog # 77063-624
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-ICAM2 Rabbit Polyclonal Antibody
Catalog # 77063-624
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A101039-100

Specifications

  • Size:
    100 µg
  • Cat. No.:
    77063-624

Specifications

About this item

Rabbit polyclonal antibody to ICAM2 for WB and ELISA with samples derived from Human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: ICAM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human