Anti-OR5M1 + OR5M10 Rabbit Polyclonal Antibody
77065-362
- Antibody Type:Primary
- Antigen Name:olfactory receptor 5M1
- Antigen Symbol:OR5M1 + OR5M10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q8NGP8
- Antigen Synonyms:OR11-208|OR11-207|OST050|Olfactory receptor family 5 subfamily M member 1|seven transmembrane helix receptor|OR5M1 + OR5M10|OR5M1|Olfactory receptor family 5 subfamily M member 10|OR11-2078
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:35 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human OR5M1 (amino acids 240 - 289)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77065-362
- Packaging:Plastic vial
Rabbit polyclonal antibody to OR5M1 for IF and ELISA with samples derived from Human.
- Validated Applications: IF, ELISA
- Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:1000
Type: Primary
Antigen: OR5M1 + OR5M10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human