Anti-ZNF75 Rabbit Polyclonal Antibody
77065-784
- Antibody Type:Primary
- Antigen Name:D8C6
- Antigen Symbol:ZNF75
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P51815
- Antigen Synonyms:ZNF75|RP13 210D15.7|Zinc finger protein 82|MGC126327|ZN75D_HUMAN|OTTHUMP00000024073|Zinc finger protein 75|Zinc finger protein 75D|ZNF75D
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:33 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human ZNF75 (amino acids 296 - 345)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77065-784
- Packaging:Plastic vial
Rabbit polyclonal antibody to ZNF75 for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: ZNF75
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human