Anti-ATP5L2 + ATP5L [EPR15636] Rabbit Polyclonal Antibody
Catalog # 77070-174
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:ATP synthase subunit g 2, mitochondrial
- Antigen Symbol:ATP5L2 + ATP5L [EPR15636]
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q7Z4Y8
- Antigen Synonyms:ATP5L|ATP5L2 + ATP5L [EPR15636]|ATPase subunit g|ATP5L2|ATP synthase subunit g, mitochondrial|ATP5K2|ATPase subunit g 2
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:11 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human ATP5L2 (amino acids 50 - 99)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77070-174
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to ATP5L2 for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: ATP5L2 + ATP5L [EPR15636]
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human