Anti-5HT1E Receptor Rabbit Polyclonal Antibody
Catalog # 77062-878
Supplier: Antibodies.com
Specifications
- Antibody Type:Primary
- Antigen Name:5 HT1E
- Antigen Symbol:5HT1E Receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P28566
- Antigen Synonyms:Serotonin receptor 1E|S31|5 hydroxytryptamine (serotonin) receptor 1E|5 hydroxytryptamine 1E receptor|5-hydroxytryptamine receptor 1E|5 hydroxytryptamine (serotonin) receptor 1E G protein coupled|5-HT-1E|HTR1E|5HT1E Receptor
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:41 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human 5 - HT - 1E (amino acids 101 - 150)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77062-878
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to 5-HT-1E for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:1000
Type: Primary
Antigen: 5HT1E Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human