Anti-CD26 Rabbit Polyclonal Antibody
Catalog # 77069-954
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:ADA-binding protein
- Antigen Symbol:CD26
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P27487
- Antigen Synonyms:ADCP 2|CD26 antigen 3|ADCP2|Adenosine deaminase complexing protein 2|CD 26|ADABP|CD26 antigen|ADCP-2|CD26
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:88 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from the Internal region of human DPP4 (amino acids 350 - 400)
- Tested Applications:IHC
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
- Cat. No.:77069-954
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to DPP4 for WB, IHC and ELISA with samples derived from Human.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500 to 1:2000, IHC-P: 1:100 to 1:300, ELISA: 1:10000
Type: Primary
Antigen: CD26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human