Anti-OR4F4 + 4F5 + 4F17 Rabbit Polyclonal Antibody
Catalog # 77073-694
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:HS14a-1-A
- Antigen Symbol:OR4F4 + 4F5 + 4F17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q96R69
- Antigen Synonyms:Olfactory receptor 4F5|Olfactory receptor 4F11|Olfactory receptor 4F18|Olfactory receptor 4F4|Olfactory receptor family 4 subfamily F member 17|Olfactory receptor family 4 subfamily F member 11 pseudogene|OLA-7501|Olfactory receptor 4F19|Olfactory receptor 4F17
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:34 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human OR4F4 (amino acids 256 - 305)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77073-694
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to OR4F4 for WB, IF and ELISA with samples derived from Human.
- Validated Applications: WB, IF, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:20000
Type: Primary
Antigen: OR4F4 + 4F5 + 4F17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human