Anti-OR5I1 Rabbit Polyclonal Antibody
77081-362
- Antibody Type:Primary
- Antigen Name:HSOlf1
- Antigen Symbol:OR5I1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q13606
- Antigen Synonyms:Olfactory receptor 5I1|Olfactory receptor-like protein OLF1|Olfactory receptor, family 5, subfamily I, member 1|Olfactory receptor, family 5, subfamily1, member 1|Olfactory receptor OR11-159|OR5I1|OR5I1_HUMAN
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:36 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human OR5I1 (amino acids 206 - 255)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77081-362
- Packaging:Plastic vial
Rabbit polyclonal antibody to OR5I1 for WB, IF and ELISA with samples derived from Human.
- Validated Applications: WB, IF, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:10000
Type: Primary
Antigen: OR5I1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human