To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CD 264
- Antigen Symbol:DcR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q9UBN6
- Antigen Synonyms:TNF receptor related receptor for TRAIL|TNF related apoptosis inducing ligand receptor 4|DcR 2|Decoy with truncated death domain|DcR2|OTTHUMP00000123528|CD264 antigen|CD264|Decoy receptor 2
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:41 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human TNFRSF10D (amino acids 121 - 170)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77076-256
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to TNFRSF10D for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: DcR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human