To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Apolipoprotein L-II
- Antigen Symbol:Apolipoprotein L 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q9BQE5
- Antigen Synonyms:APOL2_HUMAN|APOL2|Apolipoprotein L, 2|Apol8|Apolipoprotein L 2|ApoL-II|9830006J20Rik|ApoLII|APOL3
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:37 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human APOL2 (amino acids 180 - 229)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77069-168
- Packaging:Plastic vial
Rabbit polyclonal antibody to APOL2 for WB, IHC and ELISA with samples derived from Human.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: Apolipoprotein L 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human