Anti-Glucose 6 Phosphate Isomerase Rabbit Monoclonal Antibody [Clone: ARC0997]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC0997] antibody to Glucose 6 Phosphate Isomerase for WB with samples derived from Human and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: Glucose 6 Phosphate Isomerase
Clonality: Monoclonal
Clone: ARC0997
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat