Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-SARS-CoV-2 Nucleocapsid Protein Rabbit Recombinant Antibody [Clone: RMH03]
Anti-SARS-CoV-2 Nucleocapsid Protein Rabbit Recombinant Antibody [Clone: RMH03]
Catalog # 76835-024
Supplier:  Antibodies.com
Anti-SARS-CoV-2 Nucleocapsid Protein Rabbit Recombinant Antibody [Clone: RMH03]
Catalog # 76835-024
Supplier:  Antibodies.com
Supplier Number:  A290887-100

Specifications

  • Cat. No.:
    76835-024

Specifications

About this item

Recombinant rabbit monoclonal [RMH03] antibody to SARS-CoV-2 Nucleocapsid Protein for ELISA with samples derived from SARS-CoV-2.

  • Validated Applications: ELISA

Type: Primary
Antigen: SARS-CoV-2 Nucleocapsid Protein
Clonality: Recombinant
Clone: RMH03
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: SARS-CoV-2