Anti-CD51/CD61 Mouse Monoclonal Antibody [Clone: 23C6] (PE (Phycoerythrin))
Catalog # 76964-118
Supplier: Antibodies.com
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD51/CD61
- Clonality:Monoclonal
- Clone:23C6
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Chicken,Human
- Size:100 T
- Form:Liquid
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 15 mM Sodium Azide
- Storage Temperature:Store in the dark at 2...8 °C. Avoid prolonged exposure to light. Do not freeze!
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:A cell suspension containing osteoclasts from osteoclastomas.
- Purification:This antibody is conjugated with PE under optimum conditions The conjugate is purified by size-exclusion chromatography
- Cat. No.:76964-118
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [23C6] antibody to CD51/CD61 (PE) for Flow Cytometry with samples derived from Human and Chicken.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: The reagent is designed for analysis of human blood cells using 10 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD51/CD61
Clonality: Monoclonal
Clone: 23C6
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Chicken