Anti-Integrin beta 3 Mouse Monoclonal Antibody [Clone: VIPL2] (PerCP-Cyanine 5.5)
Catalog # 76954-468
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:BDPLT16
- Antigen Symbol:Integrin beta 3
- Clonality:Monoclonal
- Clone:VIPL2
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Non-human primates,Human
- Size:100 T
- Form:Liquid
- Gene ID:UniprotID# P05106
- Antigen Synonyms:HPA 1|CD61 antigen|GP3A|BDPLT2|CD 61|HPA 4|GPIIIa|GT|CD61
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 15 mM Sodium Azide
- Storage Temperature:Store in the dark at 2...8 °C. Avoid prolonged exposure to light. Do not freeze!
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Unfortunately, this information is considered to be commercially sensitive.
- Purification:This antibody is conjugated with PerCP-Cyanine 55 under optimum conditions The conjugate is purified by size-exclusion chromatography
- Cat. No.:76954-468
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [VIPL2] antibody to Integrin beta 3 (PerCP-Cyanine 5.5) for Flow Cytometry with samples derived from Human and Non-Human Primates.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: The reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: Integrin beta 3
Clonality: Monoclonal
Clone: VIPL2
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Non-Human Primates